Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039304.hg.1 |
TC01002526.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039304.hg.1 |
ENST00000373036.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039304.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039302.hg.1 |
TC01002526.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039302.hg.1 |
ENST00000373036.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039302.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039302.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013757.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202704.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202704.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202704.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202704.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202704.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202704.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202704.hg.1 |
TC0100013757.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100202707.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202707.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202707.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202707.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202707.hg.1 |
TC0100013757.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100202706.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202706.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202706.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202706.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202706.hg.1 |
TC0100013757.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100202703.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202703.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202703.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202703.hg.1 |
ENST00000373036.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202703.hg.1 |
ENST00000468190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202703.hg.1 |
NM_005955.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202703.hg.1 |
TC0100013757.hg.1 |
Clariom D TC |