Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005017.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005017.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005017.hg.1 |
TC10000428.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007925.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153777.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153777.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153777.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153777.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153777.hg.1 |
TC1000007925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000153776.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153776.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153776.hg.1 |
ENST00000373218.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153776.hg.1 |
NM_004096.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153776.hg.1 |
TC1000007925.hg.1 |
Clariom D TC |