Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027219.hg.1 |
TC05001954.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027219.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027219.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
ENST00000520200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012514.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187343.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187343.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187343.hg.1 |
ENST00000261800.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500187343.hg.1 |
NM_001447.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500187343.hg.1 |
TC0500012514.hg.1 |
Clariom D TC |