Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000565808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000565808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000565808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008926.hg.1 |
ENST00000299642.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008926.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008926.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008926.hg.1 |
TC16000630.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
ENST00000565808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
ENST00000565808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600163643.hg.1 |
TC1600008504.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000299642.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000565808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000565808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000565808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
ENST00000567430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_001244755.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008504.hg.1 |
NM_005752.4 |
Refseq |