Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009656.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009656.hg.1 |
ENST00000403586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009656.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22009656.hg.1 |
TC22000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000426145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000426145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
ENST00000426145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000426145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000426145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
ENST00000426145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008069.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157726.hg.1 |
TC2200008069.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200157726.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157726.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157726.hg.1 |
NM_004173.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200157726.hg.1 |
ENST00000382932.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157726.hg.1 |
ENST00000403586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200157726.hg.1 |
NM_004173.2 |
Refseq |