Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013925.hg.1 |
ENST00000225665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013925.hg.1 |
ENST00000544061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013925.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013925.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013925.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013925.hg.1 |
TC17001035.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000570543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000570543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000570543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000570543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173409.hg.1 |
TC1700009538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000225665.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000544061.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000570543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000570543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000570543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000570543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000574710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
ENST00000576951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_001165418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009538.hg.1 |
NM_003562.4 |
Refseq |