Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
ENST00000327238.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
ENST00000327253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
ENST00000327277.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
ENST00000355839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
ENST00000370782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
ENST00000422291.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
ENST00000438925.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
NM_022362.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020042.hg.1 |
TC10001567.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000327238.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000355839.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000370782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000415383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000434538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000438925.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000485400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000495415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
NM_001289403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
NM_001289404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
NM_001289405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
NM_022362.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000327238.14 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000355839.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000370782.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000415383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000434538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000438925.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000485400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
ENST00000495415.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
NM_001289403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
NM_001289404.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
NM_001289405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
NM_022362.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178648.hg.1 |
TC1000011553.hg.1 |
Clariom D TC |