Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016067.hg.1 |
ENST00000245138.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016067.hg.1 |
ENST00000317147.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016067.hg.1 |
ENST00000394200.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016067.hg.1 |
ENST00000441024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016067.hg.1 |
NM_016284.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016067.hg.1 |
NM_206999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016067.hg.1 |
TC16001154.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000317147.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000441024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000567188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000567285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000569240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
NM_001265612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
NM_016284.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
NM_206999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
NR_049763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000317147.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000441024.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000567188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000567285.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
ENST00000569240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
NM_001265612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
NM_016284.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
NM_206999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
NR_049763.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600180666.hg.1 |
TC1600010490.hg.1 |
Clariom D TC |