Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
TC09000647.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
ENST00000259460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
ENST00000373538.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
ENST00000373544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
ENST00000394124.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
ENST00000394125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007431.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008748.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000259460.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000373538.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000373544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000394125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000416065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
ENST00000628863.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
NM_001174152.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
NM_001174153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
NM_005833.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158324.hg.1 |
TC0900008748.hg.1 |
Clariom D TC |