Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021201.hg.1 |
TC01001351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021201.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021201.hg.1 |
ENST00000368115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021201.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010252.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175245.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175245.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175245.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175245.hg.1 |
ENST00000368114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175245.hg.1 |
ENST00000368115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175245.hg.1 |
NM_002001.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175245.hg.1 |
TC0100010252.hg.1 |
Clariom D TC |