Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000438570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000438570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000438570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000468008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000468008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
ENST00000468008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008377.hg.1 |
TC02000539.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008377.hg.1 |
ENST00000295833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008377.hg.1 |
ENST00000419482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008377.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008377.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157952.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157952.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157952.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157952.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157952.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200157952.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200157952.hg.1 |
TC0200008334.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000419482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000438570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000438570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000438570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000444564.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000468008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000468008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
ENST00000468008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008334.hg.1 |
NM_198274.3 |
Refseq |