Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006868.hg.1 |
TC01000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006868.hg.1 |
ENST00000326544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006868.hg.1 |
ENST00000361328.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006868.hg.1 |
ENST00000401072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006868.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006868.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000373413.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007725.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154390.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154390.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154390.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154390.hg.1 |
ENST00000361328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154390.hg.1 |
ENST00000480917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154390.hg.1 |
NM_145238.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154390.hg.1 |
TC0100007725.hg.1 |
Clariom D TC |