Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000285670.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000400566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000463807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000463807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000463807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000493640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000493640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000493640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000493640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
ENST00000619120.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001256370.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_001286523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007599.hg.1 |
NM_022136.4 |
Refseq |