Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036819.hg.1 |
TC02002289.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036819.hg.1 |
ENST00000272647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036819.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036819.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036819.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203718.hg.1 |
TC0200014190.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000272647.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
ENST00000393001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_001199140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014190.hg.1 |
NM_031445.2 |
Refseq |