Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010528.hg.1 |
ENST00000256545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010528.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010528.hg.1 |
TC15001190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010530.hg.1 |
ENST00000256545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010530.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010530.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010530.hg.1 |
TC15001190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164744.hg.1 |
TC1500008978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500164742.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164742.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164742.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164742.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164742.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164742.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164742.hg.1 |
TC1500008978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164745.hg.1 |
TC1500008978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500164749.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164749.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164749.hg.1 |
TC1500008978.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000256545.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000527822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000528949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
ENST00000532113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008978.hg.1 |
NM_020154.2 |
Refseq |