Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278913.1 | NP_001265842.1 | 3 - 4 | 562 | 94 | |
XM_005250187.1 | XP_005250244.1 | 3 - 4 | 239 | 94 | ||
XM_011515870.1 | XP_011514172.1 | 3 - 4 | 579 | 94 | ||
GenBank mRNA | AK075554.1 | - | 3 - 4 | 245 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |