Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000265753.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000353999.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000479338.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000489484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
ENST00000495187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_022170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007998.hg.1 |
NM_031992.1 |
Refseq |