Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014009.3 | NP_054728.2 | 3 - 4 | 503 | 55 | |
XM_006724533.2 | XP_006724596.2 | 3 - 4 | 609 | 55 | ||
XM_017029565.1 | XP_016885054.1 | 4 - 5 | 1964 | 55 | ||
GenBank mRNA | AF277993.1 | - | 3 - 4 | 503 | 55 | |
AK292052.1 | - | 3 - 4 | 387 | 55 | ||
BC113401.1 | - | 2 - 3 | 345 | 8322453 | 55 | |
BC113403.1 | - | 2 - 3 | 345 | 8322455 | 55 | |
BC143786.1 | - | 2 - 3 | 345 | 9052300 | 55 | |
EF534714.1 | - | 2 - 3 | 315 | 55 | ||
EU855812.1 | - | 2 - 3 | 315 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |