Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026278.hg.1 |
ENST00000269385.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026278.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026278.hg.1 |
TC17001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026277.hg.1 |
ENST00000269385.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026277.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026277.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026277.hg.1 |
TC17001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026280.hg.1 |
ENST00000269385.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026280.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026280.hg.1 |
TC17001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026281.hg.1 |
ENST00000269385.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026281.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026281.hg.1 |
TC17001929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026279.hg.1 |
ENST00000269385.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026279.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026279.hg.1 |
TC17001929.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700197601.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197601.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197601.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197601.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197601.hg.1 |
TC1700011949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197597.hg.1 |
TC1700011949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197598.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197598.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197598.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197598.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197598.hg.1 |
TC1700011949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011949.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
ENST00000413392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
ENST00000427800.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
ENST00000485449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197596.hg.1 |
TC1700011949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197600.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197600.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197600.hg.1 |
ENST00000269385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197600.hg.1 |
NM_020649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197600.hg.1 |
TC1700011949.hg.1 |
Clariom D TC |