Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015504.hg.1 |
ENST00000354629.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015504.hg.1 |
ENST00000360345.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015504.hg.1 |
ENST00000540085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015504.hg.1 |
ENST00000542353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015504.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015504.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015504.hg.1 |
TC12001165.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173573.hg.1 |
TC1200009793.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000354629.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000360345.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000537530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000540085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000542353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
ENST00000543765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_130441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009793.hg.1 |
NM_203503.1 |
Refseq |