Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000210313.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373904.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373920.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373920.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373920.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000373920.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000471789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000471789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000471789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000471789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000471789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000476949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000476949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000476949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000476949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000476949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000476949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000496688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000496688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000496688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000496688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000625444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000625444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
ENST00000625444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_001270427.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011385.hg.1 |
NM_005047.3 |
Refseq |