Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000258449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000393359.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
ENST00000595531.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_001142621.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013746.hg.1 |
NM_004257.5 |
Refseq |