Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000390888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004448.hg.1 |
ENST00000321990.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004448.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004448.hg.1 |
TC17000355.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700153480.hg.1 |
TC1700007469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000321990.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000390888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000578295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
ENST00000585133.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007469.hg.1 |
NM_024857.4 |
Refseq |