Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011942.hg.1 |
TC02000760.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011942.hg.1 |
ENST00000272519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011942.hg.1 |
ENST00000404963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011942.hg.1 |
ENST00000420510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011942.hg.1 |
ENST00000474855.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011942.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011945.hg.1 |
TC02000760.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011945.hg.1 |
ENST00000272519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011945.hg.1 |
ENST00000404963.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011945.hg.1 |
ENST00000420510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011945.hg.1 |
ENST00000474855.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02011945.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163790.hg.1 |
TC0200009091.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000470417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
ENST00000631312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009091.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000272519.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000412383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000420510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000431732.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000447591.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
ENST00000449649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
NM_002881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200163794.hg.1 |
TC0200009091.hg.1 |
Clariom D TC |