Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000318724.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000318724.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000318724.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000318724.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000318724.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000318724.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000318724.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000419735.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000464874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000464874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000464874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000464874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000472910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000474433.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000477775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000478915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000478915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000478915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000478915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000481880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000481880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000481880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000481880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000481880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000484475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000484475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000484475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000484475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000484475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000484475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000484475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000485652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000485652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000485652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000485652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
ENST00000485652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_020725.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_138495.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_152749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_152749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_152749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_152749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_152749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_152749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_152749.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012165.hg.1 |
NM_152749.3 |
Refseq |