Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286561.1 | NP_001273490.1 | 25 - 26 | 6516 | 86 | |
NM_006185.3 | NP_006176.2 | 25 - 26 | 6460 | 86 | ||
NR_104476.1 | 23 - 24 | 2971 | 86 | |||
XM_006718564.1 | XP_006718627.1 | 24 - 25 | 6275 | 86 | ||
XM_011545054.1 | XP_011543356.1 | 25 - 26 | 6329 | 86 | ||
XM_011545055.1 | XP_011543357.1 | 26 - 27 | 6507 | 86 | ||
XM_011545056.2 | XP_011543358.1 | 27 - 28 | 6501 | 86 | ||
XM_011545057.2 | XP_011543359.1 | 27 - 28 | 6491 | 86 | ||
XM_011545058.1 | XP_011543360.1 | 25 - 26 | 6528 | 86 | ||
XM_011545059.2 | XP_011543361.1 | 27 - 28 | 6493 | 86 | ||
XM_011545060.2 | XP_011543362.1 | 27 - 28 | 6487 | 86 | ||
XM_011545061.2 | XP_011543363.1 | 25 - 26 | 6347 | 86 | ||
XM_011545062.2 | XP_011543364.1 | 25 - 26 | 6360 | 86 | ||
XM_011545063.2 | XP_011543365.1 | 25 - 26 | 6327 | 86 | ||
XM_011545064.1 | XP_011543366.1 | 25 - 26 | 6314 | 86 | ||
XM_011545065.1 | XP_011543367.1 | 24 - 25 | 6287 | 86 | ||
XM_011545066.1 | XP_011543368.1 | 23 - 24 | 6233 | 86 | ||
XM_017017830.1 | XP_016873319.1 | 24 - 25 | 6273 | 86 | ||
XM_017017831.1 | XP_016873320.1 | 23 - 24 | 6269 | 86 | ||
GenBank mRNA | AB208841.1 | - | 10 - 11 | 4533 | 86 | |
AB210007.1 | - | 25 - 26 | 6261 | 86 | ||
BC004165.2 | - | 23 - 24 | 2886 | 86 | ||
BC008345.1 | - | 11 - 12 | 2668 | 86 | ||
BC013023.2 | - | 10 - 11 | 3362 | 3506457 | 86 | |
Z11583.1 | - | 25 - 26 | 6278 | 86 | ||
Z11584.1 | - | 25 - 26 | 6338 | 86 | ||
Z14229.1 | - | 8 - 9 | 1397 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |