Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035685.hg.1 |
TC03001997.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035685.hg.1 |
ENST00000314251.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035685.hg.1 |
ENST00000382808.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035685.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000471379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000471379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000471379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035683.hg.1 |
TC03001997.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035683.hg.1 |
ENST00000314251.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035683.hg.1 |
ENST00000382808.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03035683.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000471379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000471379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000471379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013107.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198023.hg.1 |
TC0300013107.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000471379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000314251.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000461867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000469787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000471379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
ENST00000497642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
NM_000340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
NM_001278658.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
NM_001278659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198025.hg.1 |
TC0300013107.hg.1 |
Clariom D TC |