Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002718.4 | NP_002709.2 | 2 - 3 | 2610 | 76 | |
XM_006713686.3 | XP_006713749.1 | 2 - 3 | 3021 | 76 | ||
XM_011512956.2 | XP_011511258.1 | 2 - 3 | 2785 | 76 | ||
GenBank mRNA | AK293012.1 | - | 2 - 3 | 2590 | 76 | |
AK314243.1 | - | 2 - 3 | 2538 | 76 | ||
BC065531.1 | - | 2 - 3 | 2557 | 6161260 | 76 | |
L07590.1 | - | 2 - 3 | 2497 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons |