Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011540542.1 | XP_011538844.1 | 2 - 3 | 206 | 82 | |
XM_017000131.1 | XP_016855620.1 | 1 - 2 | 1537 | 82 | ||
XM_017000132.1 | XP_016855621.1 | 2 - 3 | 296 | 82 | ||
XM_017000133.1 | XP_016855622.1 | 2 - 3 | 379 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |