Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010981.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010981.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010981.hg.1 |
TC15001224.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000559547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000559547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000559692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000559692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
ENST00000559692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000559547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000559547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000559692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000559692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
ENST00000559692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010853.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500184306.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184306.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500184306.hg.1 |
ENST00000350221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500184306.hg.1 |
NM_152597.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500184306.hg.1 |
TC1500010853.hg.1 |
Clariom D TC |