Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019241.hg.1 |
TC01001262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019241.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019241.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019241.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019242.hg.1 |
TC01001262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019242.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019242.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019243.hg.1 |
TC01001262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019243.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019243.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172810.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172810.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172810.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172810.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172810.hg.1 |
TC0100010047.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100172809.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172809.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172809.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172809.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172809.hg.1 |
TC0100010047.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100172808.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172808.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172808.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172808.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172808.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172808.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172808.hg.1 |
TC0100010047.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368677.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000368680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
ENST00000413826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010047.hg.1 |
NM_000906.3 |
Refseq |