Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006432.hg.1 |
ENST00000339736.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006432.hg.1 |
ENST00000372134.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006432.hg.1 |
ENST00000436406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006432.hg.1 |
ENST00000538477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006432.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006432.hg.1 |
TC10000591.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006433.hg.1 |
ENST00000372134.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006433.hg.1 |
ENST00000436406.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006433.hg.1 |
ENST00000538477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006433.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006433.hg.1 |
TC10000591.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155991.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155991.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155991.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155991.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155991.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000155991.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000155991.hg.1 |
TC1000008269.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000372134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
ENST00000618907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008269.hg.1 |
NM_014394.2 |
Refseq |