Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037019.hg.1 |
TC01002400.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037019.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037019.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013415.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013415.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013415.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013415.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013415.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013415.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013415.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013415.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037018.hg.1 |
TC01002400.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037018.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01037018.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199649.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199649.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199649.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199649.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199649.hg.1 |
TC0100013415.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100199647.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199647.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199647.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199647.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199647.hg.1 |
TC0100013415.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013415.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013415.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013415.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013415.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013415.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013415.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013415.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013415.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199648.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199648.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199648.hg.1 |
ENST00000289166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199648.hg.1 |
NM_052943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199648.hg.1 |
TC0100013415.hg.1 |
Clariom D TC |