Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014503.2 | NP_055318.2 | 19 - 20 | 2390 | 117 | |
GenBank mRNA | AJ006778.1 | - | 19 - 20 | 2378 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |