Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005655.hg.1 |
ENST00000267103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005655.hg.1 |
ENST00000545214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005655.hg.1 |
ENST00000548632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005655.hg.1 |
ENST00000548845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005655.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005655.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005655.hg.1 |
TC12000436.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155203.hg.1 |
TC1200007710.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200155201.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155201.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155201.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155201.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155201.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155201.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155201.hg.1 |
TC1200007710.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200155200.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155200.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155200.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155200.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155200.hg.1 |
TC1200007710.hg.1 |
Clariom D TC |