Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008331.hg.1 |
TC05000602.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008331.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008331.hg.1 |
ENST00000456348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008331.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000514725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
ENST00000514725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008328.hg.1 |
TC05000602.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008328.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008328.hg.1 |
ENST00000456348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008328.hg.1 |
ENST00000514725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008328.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008329.hg.1 |
TC05000602.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008329.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008329.hg.1 |
ENST00000456348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008329.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008332.hg.1 |
TC05000602.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008332.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008332.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000513813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000514725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
ENST00000514725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008540.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158562.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158562.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158562.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158562.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158562.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158562.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158562.hg.1 |
TC0500008540.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
ENST00000297540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
ENST00000505674.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
ENST00000511371.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
NM_032177.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158564.hg.1 |
TC0500008540.hg.1 |
Clariom D TC |