Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008943.hg.1 |
TC08002626.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008943.hg.1 |
ENST00000407775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008943.hg.1 |
ENST00000520492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008943.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008942.hg.1 |
TC08002626.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008942.hg.1 |
ENST00000407775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008942.hg.1 |
ENST00000520492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08008942.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000522160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000522160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800185975.hg.1 |
TC0800012344.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000407775.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000511341.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000517361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520027.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000520492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000522296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
ENST00000524235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012344.hg.1 |
NM_012082.3 |
Refseq |