Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011641.hg.1 |
ENST00000252729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011641.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011641.hg.1 |
TC19000870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000346968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000346968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000346968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000352529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000352529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000352529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
ENST00000352529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_031897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_031897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_031897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_031897.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NR_102308.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NR_102308.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NR_102308.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008828.hg.1 |
NR_102308.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168735.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168735.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168735.hg.1 |
TC1900008828.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900168735.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168735.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000252729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000346968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000346968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000346968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000352529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000352529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000352529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
ENST00000352529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_031897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_031897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_031897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_031897.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NM_145815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NR_102308.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NR_102308.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NR_102308.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008828.hg.1 |
NR_102308.1 |
Refseq |