Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005572.hg.1 |
ENST00000316569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005572.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005572.hg.1 |
ENST00000444228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005572.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005572.hg.1 |
TC16000376.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567308.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007532.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
ENST00000389202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
ENST00000444228.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
ENST00000567545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
NM_005353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600155277.hg.1 |
TC1600007532.hg.1 |
Clariom D TC |