Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003565.hg.1 |
TC08000250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003565.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003566.hg.1 |
TC08000250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003566.hg.1 |
ENST00000520502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003566.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003567.hg.1 |
TC08000250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003567.hg.1 |
ENST00000520502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003567.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003568.hg.1 |
TC08000250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003568.hg.1 |
ENST00000520502.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003568.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149653.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149653.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149653.hg.1 |
TC0800007221.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800149654.hg.1 |
ENST00000520502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149654.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149654.hg.1 |
ENST00000520502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149654.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149654.hg.1 |
TC0800007221.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800149655.hg.1 |
ENST00000520502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149655.hg.1 |
ENST00000523041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149655.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149655.hg.1 |
ENST00000520502.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149655.hg.1 |
ENST00000523041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149655.hg.1 |
NM_013959.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149655.hg.1 |
TC0800007221.hg.1 |
Clariom D TC |