Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
TC05002110.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
ENST00000355572.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
ENST00000355841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
ENST00000356618.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
ENST00000359895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
ENST00000393551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
NM_005451.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
NM_203352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029134.hg.1 |
NM_213636.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
TC05002110.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
ENST00000355572.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
ENST00000355841.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
ENST00000356618.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
ENST00000359895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
ENST00000393551.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
NM_005451.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
NM_203352.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05029135.hg.1 |
NM_213636.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000355572.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000355841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000359895.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000393546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000393551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000463411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000486828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000493815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000503346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000503827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000504318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000505074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000506161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000506537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
NM_005451.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
NM_203352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
NM_213636.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
NR_103804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000355572.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000355841.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000359895.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000393546.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000393551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000463411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000486828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000493815.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000503346.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000503827.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000504318.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000505074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000506161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
ENST00000506537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
NM_005451.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
NM_203352.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
NM_213636.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
NR_103804.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190536.hg.1 |
TC0500012977.hg.1 |
Clariom D TC |