Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009452.hg.1 |
TC09000744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009452.hg.1 |
ENST00000355048.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009452.hg.1 |
ENST00000361069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009452.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000355452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000462567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
ENST00000480883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008980.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161050.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161050.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161050.hg.1 |
ENST00000361069.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161050.hg.1 |
NM_006059.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161050.hg.1 |
TC0900008980.hg.1 |
Clariom D TC |