Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015463.hg.1 |
TC08001204.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015463.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015463.hg.1 |
ENST00000543296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015463.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000519183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000519183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000519183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000519183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170659.hg.1 |
TC0800010410.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000356297.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000519183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000519183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000519183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000519183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
ENST00000522933.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010410.hg.1 |
NM_144651.4 |
Refseq |