Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000310157.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376851.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000376853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000391808.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000594641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000597379.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
ENST00000599881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012964.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_001012965.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011229.hg.1 |
NM_002774.3 |
Refseq |