Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000459954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000459954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000459954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000459954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221017.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221017.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221017.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221017.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221017.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221017.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221017.hg.1 |
TC0100015984.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000368309.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000459954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000459954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000459954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000459954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
ENST00000472638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_001304342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015984.hg.1 |
NM_020131.4 |
Refseq |