Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283041.1 | NP_001269970.1 | 1 - 2 | 414 | 100 | |
NM_001283042.1 | NP_001269971.1 | 1 - 2 | 414 | 100 | ||
NM_013396.4 | NP_037528.3 | 1 - 2 | 414 | 100 | ||
XM_011529529.1 | XP_011527831.1 | 1 - 2 | 492 | 100 | ||
XM_011529535.2 | XP_011527837.1 | 1 - 2 | 492 | 100 | ||
XM_017028322.1 | XP_016883811.1 | 1 - 2 | 492 | 100 | ||
XM_017028325.1 | XP_016883814.1 | 1 - 2 | 492 | 100 | ||
XM_017028328.1 | XP_016883817.1 | 1 - 2 | 492 | 100 | ||
GenBank mRNA | AF134213.1 | - | 1 - 2 | 244 | 100 | |
AF170562.1 | - | 1 - 2 | 414 | 100 | ||
BC015930.1 | - | 1 - 2 | 104 | 100 | ||
BC075792.1 | - | 1 - 2 | 415 | 30346400 | 100 | |
FJ763652.1 | - | 1 - 2 | 45 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |