Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005255.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005255.hg.1 |
ENST00000395019.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005255.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005255.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005255.hg.1 |
TC16000361.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005253.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005253.hg.1 |
ENST00000395019.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005253.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005253.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005253.hg.1 |
TC16000361.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005252.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005252.hg.1 |
ENST00000395019.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005252.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005252.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005252.hg.1 |
TC16000361.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154793.hg.1 |
TC1600007499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154791.hg.1 |
TC1600007499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600154789.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154789.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154789.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154789.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154789.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154789.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154789.hg.1 |
TC1600007499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000279804.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
ENST00000395019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001142544.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007499.hg.1 |
NM_001330.3 |
Refseq |