Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000531697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000531697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000531697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000531697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000560330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000560330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
ENST00000560330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017785.hg.1 |
ENST00000300055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017785.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017785.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017785.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017785.hg.1 |
TC15001834.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178352.hg.1 |
TC1500010399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000300055.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000430628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000531697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000531697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000531697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000531697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000560330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000560330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
ENST00000560330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_001145311.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010399.hg.1 |
NM_002666.4 |
Refseq |