Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000256857.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000256857.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000256857.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000256857.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000256857.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000256857.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000256857.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000256857.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000420468.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000420468.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000420468.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000420468.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000420468.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000420468.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000456142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000456142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000456142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000456142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000456142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000456142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000529320.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000529320.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000529320.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000529320.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000529320.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000529320.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000529320.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000529320.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000530323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000530323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000530323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
ENST00000530323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012512.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_001012513.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_002091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_002091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_002091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_002091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_002091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_002091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_002091.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007448.hg.1 |
NM_002091.4 |
Refseq |