Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022410.hg.1 |
TC07001496.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022410.hg.1 |
ENST00000333149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022410.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022410.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022411.hg.1 |
TC07001496.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022411.hg.1 |
ENST00000333149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022411.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022411.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000488217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000488217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000488217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000493498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000493498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000493498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
ENST00000493498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
ENST00000488217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
ENST00000488217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181486.hg.1 |
TC0700011489.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181485.hg.1 |
TC0700011489.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000333149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000453152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000488217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000488217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000488217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000493498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000493498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000493498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
ENST00000493498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_001281451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011489.hg.1 |
NM_178125.3 |
Refseq |